| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177834.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177834.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177835.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177835.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177836.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177836.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177837.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177837.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177838.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177838.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177839.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177839.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177840.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_001177840.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_145533.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
NM_145533.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016063.mm.1 |
TC0200002018.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177833.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177833.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177834.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177834.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177835.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177835.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177836.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177836.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177837.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177837.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177838.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177838.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177839.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177839.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177840.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_001177840.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_145533.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
NM_145533.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200016060.mm.1 |
TC0200002018.mm.1 |
MTA TC |