Assay Details
| Gene Symbol: Bckdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009739.3 | NP_033869.1 | 4 - 5 | 641 | 90 | |
| GenBank mRNA | AF043070.1 | - | 3 - 4 | 482 | 90 | |
| AK075602.1 | - | 4 - 5 | 567 | 90 | ||
| AK145479.1 | - | 4 - 5 | 612 | 90 | ||
| AK150607.1 | - | 4 - 5 | 633 | 90 | ||
| AK150723.1 | - | 4 - 5 | 633 | 90 | ||
| AK167519.1 | - | 4 - 5 | 633 | 90 | ||
| BC004077.1 | - | 1 - 2 | 69 | 90 | ||
| BC046595.1 | - | 4 - 5 | 611 | 6313106 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bckdk
Genomic Map