Assay Details
| Gene Symbol: Sigirr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023059.3 | NP_075546.2 | 4 - 5 | 930 | 64 | |
| GenBank mRNA | AF239957.1 | - | 3 - 4 | 441 | 64 | |
| AK133518.1 | - | 4 - 5 | 555 | 64 | ||
| AK145889.1 | - | 4 - 5 | 933 | 64 | ||
| BC010806.1 | - | 3 - 4 | 452 | 4240425 | 64 | |
| BC094069.1 | - | 4 - 5 | 536 | 4974962 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sigirr
Genomic Map