| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700038400.mm.1 |
NM_028308.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038400.mm.1 |
NM_028308.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038400.mm.1 |
TC0700004550.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038395.mm.1 |
NM_028308.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038395.mm.1 |
NM_028308.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038395.mm.1 |
TC0700004550.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038398.mm.1 |
NM_028308.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038398.mm.1 |
NM_028308.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038398.mm.1 |
TC0700004550.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700038397.mm.1 |
NM_028308.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038397.mm.1 |
NM_028308.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700038397.mm.1 |
TC0700004550.mm.1 |
MTA TC |