Assay Details
| Gene Symbol: Bub3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009774.3 | NP_033904.2 | 7 - 8 | 1039 | 108 | |
| GenBank mRNA | AF149822.1 | - | 6 - 7 | 975 | 108 | |
| AK012744.1 | - | 2 - 3 | 243 | 108 | ||
| AK088534.1 | - | 7 - 8 | 1040 | 108 | ||
| BC025089.1 | - | 7 - 8 | 1051 | 3586753 | 108 | |
| BC086790.1 | - | 5 - 6 | 3004 | 108 | ||
| U67327.1 | - | 7 - 8 | 1026 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381873 | BC025089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381873 | BC086790.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381873 | U67327.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381873 | AK088534.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381873 | AF149822.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381873 | BC025089.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381873 | BC086790.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381873 | U67327.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381873 | AK088534.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P381873 | AF149822.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 754145 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390918 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781792 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 876258 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 338246 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 347394 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 248084 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12499 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381873 | BC025089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381873 | BC086790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381873 | U67327.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381873 | AK088534.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381873 | AF149822.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381873 | BC025089.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381873 | BC086790.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381873 | U67327.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381873 | AK088534.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P381873 | AF149822.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381873 | BC025089.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381873 | BC086790.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381873 | U67327.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381873 | AK088534.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381873 | AF149822.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381873 | BC025089.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381873 | BC086790.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381873 | U67327.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381873 | AK088534.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P381873 | AF149822.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P110040 | BC086790.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P110040 | AK088534.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P110040 | AK012744.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P110040 | NM_009774.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P1052476 | AK083742.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P470832 | BC086790.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06372132 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06372114 | - | - |
| Whole Mouse Genome G4122A | A_52_P470832 | BC086790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P1052476 | AK083742.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381873 | BC025089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381873 | BC086790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381873 | U67327.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381873 | AK088534.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381873 | AF149822.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381873 | BC025089.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381873 | BC086790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381873 | U67327.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381873 | AK088534.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P381873 | AF149822.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P110040 | BC086790.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P110040 | AK088534.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P110040 | AK012744.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P110040 | NM_009774.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115658 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06372116 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06411090 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06411084 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06233075 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06233072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25529195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06652986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06597665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06331275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05935033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06585172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06465569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06652982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06237180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06554002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25529171 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10558250 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558251 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558253 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558256 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558255 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558252 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10558249 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1416815_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459918_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448058_PM_s_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459104_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448473_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10558248 | - | - |
| Mouse Genome 430 2.0 Array | 1459104_at | - | - |
| Mouse Genome 430 2.0 Array | 1416815_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1448058_PM_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1416815_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1459918_at | - | - |
| Mouse Genome 430 2.0 Array | 1448058_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1448473_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448473_at | - | - |
| Mouse Genome 430 2.0 Array | 1459104_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459918_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12237_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12237_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700016910.mm.1 | TC0700001927.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016910.mm.1 | NM_009774.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016911.mm.1 | TC0700001927.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016911.mm.1 | NM_009774.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700016912.mm.1 | TC0700001927.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700016912.mm.1 | NM_009774.4 | Refseq |
| Mouse Expression Set 430 | 1448058_PM_s_at | - | - |
| Mouse Expression Set 430 | 1448058_s_at | - | - |
| Mouse Expression Set 430 | 1459918_at | - | - |
| Mouse Expression Set 430 | 1459918_PM_at | - | - |
| Mouse Expression Set 430 | 1448473_at | - | - |
| Mouse Expression Set 430 | 1448473_PM_at | - | - |
| Mouse Expression Set 430 | 1416815_s_at | - | - |
| Mouse Expression Set 430 | 1416815_PM_s_at | - | - |
| Mouse Expression Set 430 | 1459104_at | - | - |
| Mouse Expression Set 430 | 1459104_PM_at | - | - |
| Murine Genome U74v2 Set | 101961_at | - | - |
| Murine Genome U74v2 Set | 101961_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416815_PM_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1459918_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1459918_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448058_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448473_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1416815_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448473_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448058_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448473_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448058_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1459918_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448473_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1459918_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448058_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416815_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1416815_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459104_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459104_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106900341 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3170546 | - | - |
| MouseWG-6 v2 Expression BeadChip | 104070156 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106900341 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3170546 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 104070156 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003170546 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2655944 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254853 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1246921 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2655944 | - | - |
| mouse-6 v1.1 (Illumina) | scl31972.8_47-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106900341 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3170546 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 104070156 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31972.8_47-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106900341 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170546 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104070156 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2655944 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1246921 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2655944 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254853 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31972.8_47-S | - | - |

Bub3
Genomic Map