Assay Details
| Gene Symbol: Elp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023876.3 | NP_076365.2 | 5 - 6 | 656 | 100 | |
| GenBank mRNA | AJ276004.1 | - | 5 - 6 | 655 | 100 | |
| AK020742.1 | - | 5 - 6 | 664 | 100 | ||
| AK034040.1 | - | 5 - 6 | 661 | 100 | ||
| AK034529.1 | - | 5 - 6 | 662 | 100 | ||
| AK037104.1 | - | 5 - 6 | 680 | 100 | ||
| AK039447.1 | - | 5 - 6 | 668 | 100 | ||
| AK039758.1 | - | 5 - 6 | 657 | 100 | ||
| AK079069.1 | - | 5 - 6 | 671 | 100 | ||
| AK168964.1 | - | 5 - 6 | 720 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elp4
Genomic Map