Assay Details
| Gene Symbol: Slc1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077514.3 | NP_001070982.1 | 1 - 2 | 624 | 61 | |
| GenBank mRNA | AK134609.1 | - | 1 - 2 | 625 | 61 | |
| AK140095.1 | - | 1 - 2 | 619 | 61 | ||
| D43796.1 | - | 1 - 2 | 204 | 61 | ||
| U75372.1 | - | 1 - 2 | 637 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK134609.1, AK140095.1, D43796.1, NM_001077514.3, U75372.1 transcripts.
Set Membership: |
|

Slc1a2
Genomic Map