Assay Details
| Gene Symbol: Sirpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291019.1 | NP_001277948.1 | 7 - 8 | 1734 | 66 | |
| NM_001291020.1 | NP_001277949.1 | 8 - 9 | 1536 | 66 | ||
| NM_001291021.1 | NP_001277950.1 | 5 - 6 | 1080 | 66 | ||
| GenBank mRNA | AK159617.1 | - | 8 - 9 | 1543 | 66 | |
| CX236057.1 | - | 5 - 6 | 665 | 66 | ||
| D87968.1 | - | 7 - 8 | 1699 | 66 | ||
| Y10349.1 | - | 7 - 8 | 1345 | 66 | ||
More Information
Set Membership: |
|

Sirpa
Genomic Map