Assay Details
| Gene Symbol: Gpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145820.1 | NP_001139292.1 | 12 - 13 | 1931 | 70 | |
| NM_010274.3 | NP_034404.3 | 12 - 13 | 1806 | 70 | ||
| GenBank mRNA | AK034353.1 | - | 12 - 13 | 1715 | 70 | |
| AK144716.1 | - | 12 - 13 | 1932 | 70 | ||
| AK151189.1 | - | 12 - 13 | 1719 | 70 | ||
| AK167152.1 | - | 12 - 13 | 1744 | 70 | ||
| D50430.1 | - | 12 - 13 | 1708 | 70 | ||
| U60987.1 | - | 12 - 13 | 1742 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpd2
Genomic Map