Assay Details
| Gene Symbol: Cttn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252572.1 | NP_001239501.1 | 11 - 12 | 1192 | 79 | |
| NM_007803.5 | NP_031829.2 | 12 - 13 | 1303 | 79 | ||
| GenBank mRNA | AK084249.1 | - | 11 - 12 | 1002 | 79 | |
| AK146856.1 | - | 12 - 13 | 1110 | 79 | ||
| AK148010.1 | - | 12 - 13 | 1106 | 79 | ||
| AK165555.1 | - | 11 - 12 | 995 | 79 | ||
| AK168699.1 | - | 12 - 13 | 1104 | 79 | ||
| AK168871.1 | - | 11 - 12 | 993 | 79 | ||
| AK168945.1 | - | 11 - 12 | 1003 | 79 | ||
| BC011434.1 | - | 11 - 12 | 985 | 3981559 | 79 | |
| BC117820.1 | - | 11 - 12 | 1005 | 79 | ||
| U03184.1 | - | 12 - 13 | 1092 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cttn
Genomic Map