Assay Details
| Gene Symbol: Pnpla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_028142.1 | 2 - 3 | 369 | 80 | ||
| GenBank mRNA | BC019188.1 | - | 2 - 3 | 351 | 5027544 | 80 |
| BC044737.1 | - | 2 - 3 | 333 | 80 | ||
| BC044781.2 | - | 2 - 3 | 333 | 5151911 | 80 | |
| BC054825.1 | - | 2 - 3 | 328 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC019188.1, BC044781.2 transcripts.
Set Membership: |
|

Pnpla2
Genomic Map