Assay Details
| Gene Symbol: Rbbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015754.2 | NP_056569.2 | 3 - 4 | 331 | 77 | |
| GenBank mRNA | AK017606.1 | - | 3 - 4 | 272 | 77 | |
| AK076944.1 | - | 3 - 4 | 326 | 77 | ||
| BC011107.1 | - | 3 - 4 | 269 | 3982875 | 77 | |
| BC050201.1 | - | 3 - 4 | 267 | 30013927 | 77 | |
| BC147814.1 | - | 3 - 4 | 324 | 9087531 | 77 | |
| BC147815.1 | - | 3 - 4 | 324 | 9087532 | 77 | |
| BY256564.1 | - | 3 - 4 | 342 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbbp9
Genomic Map