Assay Details
| Gene Symbol: Igsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030691.1 | NP_109616.1 | 3 - 4 | 552 | 65 | |
| GenBank mRNA | AJ278575.1 | - | 3 - 4 | 552 | 65 | |
| AK040569.1 | - | 3 - 4 | 532 | 65 | ||
| AK151417.1 | - | 3 - 4 | 541 | 65 | ||
| AK152186.1 | - | 3 - 4 | 541 | 65 | ||
| AK156382.1 | - | 3 - 4 | 490 | 65 | ||
| AK163058.1 | - | 3 - 4 | 532 | 65 | ||
| BC057951.1 | - | 3 - 4 | 549 | 5341788 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igsf6
Genomic Map