Assay Details
| Gene Symbol: Oraov1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028184.3 | NP_082460.2 | 2 - 3 | 242 | 80 | |
| GenBank mRNA | AK008702.1 | - | 2 - 3 | 302 | 80 | |
| AK046447.1 | - | 2 - 3 | 341 | 80 | ||
| AK083244.1 | - | 2 - 3 | 299 | 80 | ||
| AK167712.1 | - | 2 - 3 | 266 | 80 | ||
| AK169310.1 | - | 2 - 3 | 266 | 80 | ||
| BC006906.1 | - | 2 - 3 | 153 | 80 | ||
| BC013564.1 | - | 2 - 3 | 242 | 3591291 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Oraov1
Genomic Map