Assay Details
| Gene Symbol: Pdilt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027943.1 | NP_082219.1 | 8 - 9 | 1341 | 114 | |
| GenBank mRNA | AK005692.1 | - | 8 - 9 | 1341 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdilt
Genomic Map