Assay Details
Gene Symbol: Fbxl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172748.2 | NP_766336.2 | 4 - 5 | 801 | 76 | |
GenBank mRNA | AK028405.1 | - | 4 - 5 | 570 | 76 | |
BC059812.1 | - | 4 - 5 | 801 | 6401846 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
