Assay Details
| Gene Symbol: Inpp5f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047166.1 | - | 1 - 2 | 44 | 71 | |
| DQ792944.1 | - | 1 - 2 | 49 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK047166.1 transcripts.
Set Membership: |
|

Inpp5f
Genomic Map