Assay Details
| Gene Symbol: Mtg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199301.2 | NP_955005.2 | 5 - 6 | 516 | 56 | |
| GenBank mRNA | AK144453.1 | - | 5 - 6 | 441 | 56 | |
| AK144625.1 | - | 5 - 6 | 474 | 56 | ||
| BC027306.1 | - | 5 - 6 | 425 | 4019717 | 56 | |
 Back To Top
 Back To TopMore Information
Updated Mapping Information
 This assay targets the same location as MTA transcript cluster(s) TC0700002004.mm.1
					            	This assay targets the same location as MTA transcript cluster(s) TC0700002004.mm.1| Set Membership: |  Ambion siRNA available  Amplicon length less than or equal to 70  Mammalian Gene Collection  Detect Genomic DNA  Mus musculus  Probe spans exons | 
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
 Back To Top
 Back To Top
 Mtg1
Mtg1 Genomic Map
Genomic Map Back To Top
 Back To Top