Assay Details
| Gene Symbol: Prss36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081374.1 | NP_001074843.2 | 13 - 14 | 2461 | 61 | |
| GenBank mRNA | AK021185.1 | - | 3 - 4 | 417 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135860 | AK021185.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135860 | BC044738.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135860 | NM_001081374.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154652 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928554 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 590495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763456 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457098 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 681290 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451584 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 905373 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 159183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94145 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135860 | AK021185.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135860 | BC044738.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135860 | NM_001081374.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208537 | AK021185.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208537 | BC044738.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208537 | NM_001081374.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135938 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P116587 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25508274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508304 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06642783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06001248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508271 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508274 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508230 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06171652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508278 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06627944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06581786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508254 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06001247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06440952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508240 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06171647 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508236 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508238 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508302 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06440936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508286 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508307 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508234 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06440944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06627950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06216883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06440927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06440926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508248 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06581779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508250 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508267 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06642781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508256 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06266724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508288 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06305005 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508298 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508281 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508296 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06642786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25508223 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10568342 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568349 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568343 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568352 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568350 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568348 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568354 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568341 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568340 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568347 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568346 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568345 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10568344 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429164_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10568339 | - | - |
| Mouse Genome 430 2.0 Array | 1429164_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1429164_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77613_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77613_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700036528.mm.1 | TC0700004347.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036532.mm.1 | TC0700004347.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036530.mm.1 | TC0700004347.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036527.mm.1 | TC0700004347.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036526.mm.1 | TC0700004347.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036529.mm.1 | TC0700004347.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700036531.mm.1 | TC0700004347.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1429164_PM_at | - | - |
| Mouse Expression Set 430 | 1429164_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429164_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429164_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 326399 | - | - |
| ABI Mouse Genome Survey Microarray | 913506 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228060 | - | - |

Prss36
Genomic Map