Assay Details
| Gene Symbol: Syce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143765.1 | NP_001137237.1 | 10 - 11 | 764 | 95 | |
| GenBank mRNA | AK016694.1 | - | 10 - 11 | 764 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syce1
Genomic Map