Assay Details
Gene Symbol: Syce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143765.1 | NP_001137237.1 | 6 - 7 | 425 | 89 | |
GenBank mRNA | AK016694.1 | - | 6 - 7 | 425 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
