Assay Details
| Gene Symbol: Syce1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001143765.1 | NP_001137237.1 | 8 - 9 | 576 | 141 | |
| GenBank mRNA | AK016694.1 | - | 8 - 9 | 576 | 141 | |
More Information
Set Membership: |
|

Syce1
Genomic Map