Assay Details
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291056.1 | NP_001277985.1 | 1 - 2 | 145 | 61 | |
NM_011428.3 | NP_035558.1 | 1 - 2 | 145 | 61 | ||
GenBank mRNA | AK078038.1 | - | 1 - 2 | 143 | 61 | |
AK138653.1 | - | 1 - 2 | 113 | 61 | ||
AK161896.1 | - | 1 - 2 | 113 | 61 | ||
AU080862.1 | - | 1 - 2 | 112 | 61 | ||
BC018249.1 | - | 1 - 2 | 33 | 4504644 | 61 | |
CB232574.1 | - | 1 - 2 | 148 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK078038.1, BC018249.1, NM_001291056.1, NM_011428.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
