Assay Details
| Gene Symbol: Rin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028724.4 | NP_083000.5 | 4 - 5 | 408 | 66 | |
| GenBank mRNA | AK150680.1 | - | 2 - 3 | 216 | 66 | |
| AK151332.1 | - | 1 - 2 | 172 | 66 | ||
| AK152893.1 | - | 2 - 3 | 216 | 66 | ||
| BC040390.1 | - | 4 - 5 | 402 | 3156682 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK150680.1, AK151332.1, AK152893.1, BC040390.1, NM_028724.4 transcripts.
Set Membership: |
|

Rin2
Genomic Map