Assay Details
| Gene Symbol: Slc24a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053195.2 | NP_444425.1 | 11 - 12 | 965 | 80 | |
| GenBank mRNA | AF314821.1 | - | 11 - 12 | 1005 | 80 | |
| BC005742.1 | - | 1 - 2 | 95 | 80 | ||
| BC017615.1 | - | 11 - 12 | 965 | 4914291 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc24a3
Genomic Map