Assay Details
| Gene Symbol: Atf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025093.2 | NP_001020264.1 | 11 - 12 | 1195 | 63 | |
| NM_001284369.1 | NP_001271298.1 | 9 - 10 | 1035 | 63 | ||
| NM_001284370.1 | NP_001271299.1 | 11 - 12 | 1195 | 63 | ||
| NM_001284371.1 | NP_001271300.1 | 10 - 11 | 1132 | 63 | ||
| NM_001284372.1 | NP_001271301.1 | 9 - 10 | 901 | 63 | ||
| NM_001284373.1 | NP_001271302.1 | 8 - 9 | 838 | 63 | ||
| NM_001284374.1 | NP_001271303.1 | 8 - 9 | 920 | 63 | ||
| NM_001284376.1 | NP_001271305.1 | 11 - 12 | 1077 | 63 | ||
| NM_009715.3 | NP_033845.1 | 10 - 11 | 1098 | 63 | ||
| GenBank mRNA | AF483482.1 | - | 7 - 8 | 772 | 63 | |
| AF483483.1 | - | 7 - 8 | 772 | 63 | ||
| AK035438.1 | - | 11 - 12 | 1151 | 63 | ||
| AK051591.1 | - | 10 - 11 | 1088 | 63 | ||
| AK139515.1 | - | 9 - 10 | 951 | 63 | ||
| AK146839.1 | - | 10 - 11 | 1044 | 63 | ||
| AY902311.1 | - | 6 - 7 | 658 | 63 | ||
| BC042210.1 | - | 8 - 9 | 871 | 4018323 | 63 | |
| BC079883.1 | - | 10 - 11 | 1020 | 6839572 | 63 | |
| BC082596.1 | - | 8 - 9 | 784 | 30649207 | 63 | |
| BC085137.1 | - | 8 - 9 | 863 | 63 | ||
| M31629.1 | - | 3 - 4 | 250 | 63 | ||
| M77167.1 | - | 5 - 6 | 544 | 63 | ||
| S76657.1 | - | 863 | 63 | |||
| S76659.1 | - | 569 | 63 | |||
More Information
Set Membership: |
|

Atf2
Genomic Map