Assay Details
| Gene Symbol: Pdk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172665.5 | NP_766253.2 | 5 - 6 | 834 | 115 | |
| GenBank mRNA | AK046805.1 | - | 5 - 6 | 835 | 115 | |
| AK153649.1 | - | 5 - 6 | 787 | 115 | ||
| BC027196.1 | - | 5 - 6 | 757 | 4458562 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdk1
Genomic Map