Assay Details
| Gene Symbol: Ccl20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159738.1 | NP_001153210.1 | 1 - 2 | 139 | 86 | |
| GenBank mRNA | AF099052.1 | - | 1 - 2 | 112 | 86 | |
| AK079862.1 | - | 1 - 2 | 140 | 86 | ||
| BC028504.1 | - | 1 - 2 | 112 | 1380543 | 86 | |
| BY026108.1 | - | 1 - 2 | 139 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccl20
Genomic Map