Assay Details
| Gene Symbol: Vmn1r226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134191.1 | NP_598952.1 | 1 | 652 | 124 | |
| GenBank mRNA | BC119429.1 | - | 1 | 650 | 124 | |
| BC119430.1 | - | 1 | 652 | 40040474 | 124 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Vmn1r226
Genomic Map