Assay Details
| Gene Symbol: Hnrnpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109913.1 | NP_001103383.1 | 13 - 14 | 1073 | 71 | |
| NM_029804.3 | NP_084080.1 | 12 - 13 | 1190 | 71 | ||
| GenBank mRNA | AK011521.1 | - | 12 - 13 | 1136 | 71 | |
| AK145343.1 | - | 13 - 14 | 1011 | 71 | ||
| AK159848.1 | - | 13 - 14 | 1005 | 71 | ||
| AK166670.1 | - | 12 - 13 | 1125 | 71 | ||
| AK168347.1 | - | 13 - 14 | 1008 | 71 | ||
| AK171892.1 | - | 13 - 14 | 1008 | 71 | ||
| AK220299.1 | - | 13 - 14 | 1033 | 71 | ||
| BC005758.1 | - | 1 - 2 | 83 | 71 | ||
| BC017539.1 | - | 13 - 14 | 1003 | 71 | ||
| BC035525.1 | - | 13 - 14 | 1008 | 71 | ||
| BC054785.1 | - | 2592 | 71 | |||
| BC065172.1 | - | 13 - 14 | 1003 | 6415170 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpm
Genomic Map