Assay Details
| Gene Symbol: Hnrnpm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109913.1 | NP_001103383.1 | 15 - 16 | 1942 | 84 | |
| NM_029804.3 | NP_084080.1 | 14 - 15 | 2059 | 84 | ||
| GenBank mRNA | AK011521.1 | - | 14 - 15 | 2005 | 84 | |
| AK133911.1 | - | 13 - 14 | 1941 | 84 | ||
| AK145343.1 | - | 15 - 16 | 1880 | 84 | ||
| AK159848.1 | - | 15 - 16 | 1874 | 84 | ||
| AK163866.1 | - | 3 - 4 | 891 | 84 | ||
| AK168347.1 | - | 15 - 16 | 1877 | 84 | ||
| AK169101.1 | - | 15 - 16 | 1876 | 84 | ||
| AK171892.1 | - | 15 - 16 | 1877 | 84 | ||
| AK178014.1 | - | 1 - 2 | 67 | 84 | ||
| AK219463.1 | - | 1 - 2 | 67 | 84 | ||
| AK220299.1 | - | 15 - 16 | 1902 | 84 | ||
| BC005758.1 | - | 3 - 4 | 952 | 84 | ||
| BC035525.1 | - | 15 - 16 | 1877 | 84 | ||
| BC054785.1 | - | 3461 | 84 | |||
| BC065172.1 | - | 15 - 16 | 1872 | 6415170 | 84 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrnpm
Genomic Map