Assay Details
| Gene Symbol: Abcc10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145140.2 | NP_660122.1 | 2 - 3 | 2299 | 95 | |
| NM_170680.2 | NP_733780.1 | 3 - 4 | 1699 | 95 | ||
| GenBank mRNA | AF406642.1 | - | 2 - 3 | 2299 | 95 | |
| AF417121.1 | - | 3 - 4 | 1680 | 95 | ||
| AK131108.1 | - | 2 - 3 | 1719 | 95 | ||
| AK133240.1 | - | 3 - 4 | 1656 | 95 | ||
| AK170539.1 | - | 3 - 4 | 1675 | 95 | ||
| AK170600.1 | - | 3 - 4 | 1673 | 95 | ||
| AK171535.1 | - | 3 - 4 | 1675 | 95 | ||
| BC060644.1 | - | 2 - 3 | 1765 | 95 | ||
More Information
Set Membership: |
|

Abcc10
Genomic Map