Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200011542.mm.1 |
NM_178890.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011542.mm.1 |
NM_178890.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011542.mm.1 |
TC0200001535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200011544.mm.1 |
NM_178890.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011544.mm.1 |
NM_178890.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011544.mm.1 |
TC0200001535.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200011543.mm.1 |
NM_178890.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011543.mm.1 |
NM_178890.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200011543.mm.1 |
TC0200001535.mm.1 |
MTA TC |