Assay Details
| Gene Symbol: Shc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199022.2 | NP_950187.1 | 9 - 10 | 2068 | 90 | |
| GenBank mRNA | AY464564.1 | - | 9 - 10 | 1307 | 90 | |
| BC060643.1 | - | 6 - 7 | 919 | 90 | ||
| BC103713.1 | - | 8 - 9 | 1375 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Shc4
Genomic Map