Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700018040.mm.1 |
TC0700002042.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700018040.mm.1 |
NM_001172064.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018040.mm.1 |
NM_001172064.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018040.mm.1 |
NM_001172065.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018040.mm.1 |
NM_001172065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018040.mm.1 |
NM_153777.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700018040.mm.1 |
NM_153777.2 |
Refseq |