Assay Details
Gene Symbol: Lrrc56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172064.1 | NP_001165535.1 | 10 - 11 | 1608 | 128 | |
NM_153777.2 | NP_722472.1 | 10 - 11 | 1194 | 128 | ||
GenBank mRNA | AK220237.1 | - | 10 - 11 | 1562 | 128 | |
BC027807.1 | - | 10 - 11 | 1204 | 5324022 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
