Assay Details
| Gene Symbol: Cracr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025103.2 | NP_001020274.2 | 8 - 9 | 1065 | 71 | |
| GenBank mRNA | BC026645.1 | - | 5 - 6 | 639 | 71 | |
| BC034124.1 | - | 2 - 3 | 261 | 71 | ||
| BC048916.1 | - | 8 - 9 | 1065 | 71 | ||
| BC098333.1 | - | 8 - 9 | 1026 | 5256663 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cracr2b
Genomic Map