Assay Details
| Gene Symbol: Tmco5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026104.4 | NP_080380.1 | 2 - 3 | 117 | 111 | |
| GenBank mRNA | AK007078.1 | - | 2 - 3 | 101 | 111 | |
| AK007251.1 | - | 2 - 3 | 115 | 111 | ||
| BC049568.1 | - | 2 - 3 | 81 | 6742928 | 111 | |
| BU936444.1 | - | 2 - 3 | 120 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007078.1, AK007251.1, BC049568.1, NM_026104.4 transcripts.
Set Membership: |
|

Tmco5
Genomic Map