Assay Details
| Gene Symbol: Frs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144939.2 | NP_659188.1 | 6 - 7 | 802 | 63 | |
| GenBank mRNA | AK077209.1 | - | 2 - 3 | 322 | 63 | |
| BC014819.1 | - | 6 - 7 | 802 | 4506982 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213685 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213686 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020736 | BC014819.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020736 | NM_144939.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2020736 | AK077209.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92514 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179512 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872533 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150592 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 48497 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111153 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020736 | BC014819.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020736 | NM_144939.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2020736 | AK077209.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P444162 | BC014819.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31213687 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03326948 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111153 | - | - |
| Whole Mouse Genome G4122A | A_51_P444162 | BC014819.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126633 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111153 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13432307 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213685 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31213686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31213806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13414055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13557378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213740 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13575498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13096604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13203869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213703 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213727 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13414044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213808 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213791 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13148159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13096605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213723 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13575497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13432307 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13414070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213816 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13144724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13410852 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213739 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213785 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213752 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13570360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13196769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13414059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213682 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213799 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213795 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13144717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13096608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13218885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13250909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13346341 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213801 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13133296 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13204189 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13103197 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213782 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13204194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213798 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213813 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213784 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13278707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213768 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213751 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213788 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213786 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13278709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213811 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213671 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13346340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213805 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213746 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13218884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13575499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213823 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31213726 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10445712 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445717 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445711 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445716 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445714 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445713 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445715 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424449_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445710 | - | - |
| Mouse Genome 430 2.0 Array | 1439277_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424449_at | - | - |
| Mouse Genome 430 2.0 Array | 1424449_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439277_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107971_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107971_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700008898.mm.1 | NM_144939.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008898.mm.1 | NM_144939.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008898.mm.1 | TC1700002793.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430A Array | 1424449_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424449_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 477069 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5890048 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5890048 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005890048 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2638186 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2638186 | - | - |
| mouse-6 v1.1 (Illumina) | scl50645.13_521-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5890048 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50645.13_521-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5890048 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2638186 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2638186 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50645.13_521-S | - | - |

Frs3
Genomic Map