Assay Details
| Gene Symbol: Gm6249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_046021.1 | 4 - 5 | 1140 | 70 | ||
| GenBank mRNA | AK034107.1 | - | 4 - 5 | 1141 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA C230079O03 gene | C230079O03Rik | C230079O03 | -336.3kb | Sense | Chr. 7 - 136332216 - 136349415 on NCBI Build GRCm38 |
| Downstream | O-6-methylguanine-DNA methyltransferase | Mgmt |
AGT, AI267024, Agat |
187.9kb | Anti-sense | Chr. 7 - 136894468 - 137128193 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm6249
Genomic Map