Assay Details
Gene Symbol: Gm15506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK052909.1 | - | 3 - 4 | 555 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 2210018M11 gene | 2210018M11Rik |
2310016L09Rik, AU019521, AU019557, Emsy |
-24.0kb | Sense | Chr. 7 - 98587137 - 98658289 on NCBI Build GRCm38 |
Downstream |
protein-kinase, interferon-inducible double stranded RNA dependent inhibitor, repressor of (P58 repressor) |
Prkrir |
2900052B10Rik, Dap4, Rpkrir |
10.0kb | Anti-sense | Chr. 7 - 98703230 - 98718062 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042680 | AK052909.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 404914 | NM_146905.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 740838 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 567205 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 508846 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 118530 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687221 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916688 | AK004909.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916688 | AK010057.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916688 | AK013122.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916688 | AK014212.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916688 | AK150410.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916688 | AK152560.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916688 | AK159784.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916688 | AK167961.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916688 | AK168632.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916688 | BC003758.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916688 | NM_008258.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 916688 | U90123.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 329352 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 25928 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673828 | AK075863.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673828 | AK158914.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673828 | BC031720.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673828 | NM_172702.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 673828 | XM_001471842.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795654 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28022 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 242075 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346845 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156996 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 77489 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45221 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171120 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042680 | AK052909.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06295297 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171120 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01586080 | - | - |
Whole Mouse Genome G4122A | A_51_P247385 | AK052909.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P247385 | BB519491.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117197 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357272 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357252 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357250 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357242 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06491057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357255 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06491062 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357269 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06579189 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357254 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357246 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357263 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357256 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06099913 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06579193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06538948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06099916 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25357236 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1445074_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1444252_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1446456_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1445074_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1446456_at | - | - |
Mouse Genome 430 2.0 Array | 1444252_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1445074_at | - | - |
Mouse Genome 430 2.0 Array | 1446456_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1444252_at | - | - |
Mouse Expression Set 430 | 1445074_PM_at | - | - |
Mouse Expression Set 430 | 1446456_at | - | - |
Mouse Expression Set 430 | 1444252_PM_at | - | - |
Mouse Expression Set 430 | 1445074_at | - | - |
Mouse Expression Set 430 | 1444252_at | - | - |
Mouse Expression Set 430 | 1446456_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444252_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445074_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445074_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1446456_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1446456_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1444252_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 106290575 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1238835 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 106290575 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106290575 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1238835 | - | - |
