Assay Details
| Gene Symbol: Gm35692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK053380.1 | - | 1 - 2 | 115 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA F630040K05 gene | F630040K05Rik | -21.6kb | Anti-sense | Chr. 17 - 45734506 - 45786357 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930542M03 gene | 4930542M03Rik | 57.8kb | Sense | Chr. 17 - 45880119 - 45881452 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm35692
Genomic Map