Assay Details
| Gene Symbol: Mep1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008585.2 | NP_032611.2 | 12 - 13 | 1838 | 110 | |
| GenBank mRNA | AK093914.1 | - | 12 - 13 | 1797 | 110 | |
| AK136455.1 | - | 2 - 3 | 496 | 110 | ||
| AK143906.1 | - | 12 - 13 | 1796 | 110 | ||
| BC015258.1 | - | 12 - 13 | 1838 | 4241252 | 110 | |
| M74897.1 | - | 12 - 13 | 1837 | 110 | ||
More Information
Set Membership: |
|

Mep1a
Genomic Map