Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200036787.mm.1 |
NM_201367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036787.mm.1 |
NM_201367.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036787.mm.1 |
TC0200004410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200036788.mm.1 |
NM_201367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036788.mm.1 |
NM_201367.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036788.mm.1 |
TC0200004410.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200036789.mm.1 |
NM_201367.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036789.mm.1 |
NM_201367.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036789.mm.1 |
TC0200004410.mm.1 |
MTA TC |