Assay Details
| Gene Symbol: Ndst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010811.2 | NP_034941.2 | 3 - 4 | 1757 | 76 | |
| GenBank mRNA | AF074925.1 | - | 1 - 2 | 1007 | 76 | |
| AK150100.1 | - | 3 - 4 | 1538 | 76 | ||
| AK170298.1 | - | 3 - 4 | 1540 | 76 | ||
| BC050829.1 | - | 3 - 4 | 1522 | 76 | ||
| BC110479.1 | - | 1 - 2 | 1155 | 40048114 | 76 | |
| BC110480.1 | - | 1 - 2 | 1155 | 40048115 | 76 | |
| U02304.1 | - | 2 - 3 | 1519 | 76 | ||
| X75885.1 | - | 1 - 2 | 1164 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndst2
Genomic Map