Assay Details
| Gene Symbol: Ndst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010811.2 | NP_034941.2 | 1 - 2 | 348 | 160 | |
| GenBank mRNA | AF520868.1 | - | 1 - 2 | 348 | 160 | |
| AI173867.1 | - | 1 - 2 | 348 | 160 | ||
| AK150100.1 | - | 1 - 2 | 129 | 160 | ||
| AK170298.1 | - | 1 - 2 | 129 | 160 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK150100.1, NM_010811.2 transcripts.
Set Membership: |
|

Ndst2
Genomic Map