Assay Details
| Gene Symbol: Far1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005686.1 | - | 5 - 6 | 419 | 110 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | parathyroid hormone | Pth | Pthp | -149.5kb | Sense | Chr. 7 - 113385576 - 113388573 on NCBI Build GRCm38 |
| Downstream | predicted gene 29507 | Gm29507 | 189.0kb | Anti-sense | Chr. 7 - 113729654 - 113751238 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Far1os
Genomic Map