Assay Details
Gene Symbol: Pax6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244200.1 | NP_001231129.1 | 1 - 2 | 144 | 62 | |
GenBank mRNA | AK047243.1 | - | 2 - 3 | 386 | 62 | |
AY064175.1 | - | 1 - 2 | 107 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY064175.1, NM_001244200.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
