Assay Details
| Gene Symbol: Igf2os | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002855.2 | 1 - 2 | 1335 | 60 | |||
| GenBank mRNA | AB030734.1 | - | LIT1673 PEG8/Igf2as |
1 - 2 | 1334 | 60 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 483 | Mir483 |
Mirn483, mir-483, mmu-mir-483 |
-4.7kb | Anti-sense | Chr. 7 - 142654924 - 142654996 on NCBI Build GRCm38 |
| Downstream | insulin II | Ins2 |
AA986540, Ins-2, InsII, Mody, Mody4, proinsulin |
13.1kb | Anti-sense | Chr. 7 - 142678656 - 142679708 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Igf2os
Genomic Map