Assay Details
| Gene Symbol: C330022C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045717.1 | 1 - 2 | 308 | 76 | ||
| GenBank mRNA | AK021219.1 | - | 1 - 2 | 309 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | urate (5-hydroxyiso-) hydrolase | Urah |
1190003J15Rik, 2810420C16Rik |
-3.9kb | Anti-sense | Chr. 7 - 140835023 - 140837968 on NCBI Build GRCm38 |
| Downstream | outer dense fiber of sperm tails 3 | Odf3 |
1700011O04Rik, SHIPPO1 |
2.2kb | Anti-sense | Chr. 7 - 140847916 - 140850925 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

C330022C24Rik
Genomic Map